Learn More
ATCC Helicobacter pylori
Catalog No. 502382853
Encompass
Helicobacter pylori strain 26695 [KE26695] is a whole-genome sequenced bacterium that was isolated in the United Kingdom from the stomach of a patient with gastritis. This bacterial culture can be used in enteric disease research.
Catalog No. 50-238-2853 Supplier ATCC Supplier No. 700392
Add to Cart
Edge
Add to Cart
Helicobacter pylori strain 26695 [KE26695] is a whole-genome sequenced bacterium that was isolated in the United Kingdom from the stomach of a patient with gastritis. This bacterial culture can be used in enteric disease research.