Learn More
Diagnostic Biosystems HELICOBACTER PYLORI Helicobacter Pylori
Catalog No. 502407497
Encompass_Preferred
This antibody reacts with the 54 kDa (flagellin) of H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.
Catalog No. 50-240-7497 Supplier Diagnostic Biosystems Supplier No. PDM559
This antibody reacts with the 54 kDa (flagellin) of H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.