Learn More
Bioss Helicobacter pylori
SDP
Supplier:  Bioss BSM72172M
Encompass_Preferred
Helicobacter pylori Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-64957