Learn More
Diagnostic Biosystems HELICOBACTER PYLORI ANTIBODY Helicobacter Pylori Antibody
Catalog No. 502407460
Encompass_Preferred
This antibody reacts with the whole H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.
Catalog No. 50-240-7460 Supplier Diagnostic Biosystems Supplier No. PDR169
This antibody reacts with the whole H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.