Learn More
StatLab Helicobacter Pylori Control Slides
Description
- Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
- Use these slides with confidence to evaluate stain quality and performance in histology laboratories
- Includes 25 Unstained Slides per pack
Specifications
Specifications
| Type | Control Slide |
| Quantity | 25 Pk. |
| Species | Human |