Learn More
Cayman Chemical HelIcobacTR pylorI NAP RabbI
SDP
Catalog No. 502947753
Encompass_Preferred
Immunogen: Proteins purified from H. pylori • Host: Rabbit • Species Reactivity: (+) H. pylori • Cross Reactivity: (+) HP-NAP • Applications: ELISA
Catalog No. 50-294-7753 Supplier Cayman Chemical Supplier No. 32345100 UG
Immunogen: Proteins purified from H. pylori • Host: Rabbit • Species Reactivity: (+) H. pylori • Cross Reactivity: (+) HP-NAP • Applications: ELISA