Learn More
Cayman Chemical HelIcobacTR pylorI OMP RabbI
SDP
Catalog No. 502947330
Encompass_Preferred
Immunogen: Proteins purified from H. pylori • Host: Rabbit • Species Reactivity: (+) H. pylori • Cross Reactivity: (+) OMP; (-) CagA, Urease, HP-NAP • Applications: ELISA
Catalog No. 50-294-7330 Supplier Cayman Chemical Supplier No. 32342100 UG
Immunogen: Proteins purified from H. pylori • Host: Rabbit • Species Reactivity: (+) H. pylori • Cross Reactivity: (+) OMP; (-) CagA, Urease, HP-NAP • Applications: ELISA