Learn More
Cayman Chemical HelIcobacTR pylorI OMP RabbI
SDP
Supplier:  Cayman Chemical 32341100 UG
Encompass_Preferred
Immunogen: Proteins purified from H. pylori • Host: Rabbit • Species Reactivity: (+) H. pylori • Cross Reactivity: (+) OMP; (-) CagA, Urease, HP-NAP • Applications: ELISA
Catalog No. 50-294-7511