Learn More
Diagnostic Biosystems HELIOCOBACTER PYLORI Heliocobacter pylori
Catalog No. 502407640
Encompass_Preferred
This antibody reacts with the whole H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.
Catalog No. 50-240-7640 Supplier Diagnostic Biosystems Supplier No. PDR053
This antibody reacts with the whole H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.