Learn More
MSC Helios Vernier Caliper, 0 in. to 72 in., 0.001 in. Graduation, Stainless Steel, 1 ea.
Catalog No. 5000516884
Encompass
This vernier caliper is a high-precision dimensional measuring tool designed for accurate length and distance measurements up to 72 in. Constructed from durable stainless steel, it is built for reliability in demanding industrial measurement applications.
  • Measuring range of 0 in. to 72 in.
  • Graduation of 0.001 in.
  • Constructed from high-quality stainless steel
Catalog No. 50-005-16884 Supplier MSC Supplier No. 06416820
This vernier caliper is a high-precision dimensional measuring tool designed for accurate length and distance measurements up to 72 in. Constructed from durable stainless steel, it is built for reliability in demanding industrial measurement applications.
  • Measuring range of 0 in. to 72 in.
  • Graduation of 0.001 in.
  • Constructed from high-quality stainless steel