Learn More
JDM LAB SOLUTIONS LLC HELIPORT SMPL THAWING STATION
Catalog No. NC1717359
Encompass
The most complete sample thawing solution available today. Featuring an expanded thawing surface for large sample arrays. Fully customizable sample arrangement for optimum functionality. And sufficient fan power to thaw everything in your freezer quickly and reproducibly. Eliminate edge effects, cold starts and reduce process variation. All with no excess input of heat, no tweaking of controls and no calibration or maintenance required
Catalog No. NC1717359 Supplier JDM LAB SOLUTIONS LLC Supplier No. 130070000
The most complete sample thawing solution available today. Featuring an expanded thawing surface for large sample arrays. Fully customizable sample arrangement for optimum functionality. And sufficient fan power to thaw everything in your freezer quickly and reproducibly. Eliminate edge effects, cold starts and reduce process variation. All with no excess input of heat, no tweaking of controls and no calibration or maintenance required