Learn More
AdipoGen Heliquinomycin (500 µg)
SDP
Supplier:  AdipoGen AGCN20091C500
Encompass_Preferred
Chemical. CAS: 178182-49-5. Formula: C33H30O17. MW: 698.6. Isolated from Streptomyces sp. MJ929-SF2. Antibiotic. DNA helicase inhibitor. Inhibits the DNA helicase activity of the human minichromosome maintenance (MCM) 4/6/7 complex. Topoisomerase I and II inhibitor (weaker than DNA helicase). DNA and RNA synthesis inhibitor in cell culture. Does not inhibit protein synthesis. Induces cell cycle arrest. Anticancer compound. Antibacterial against Gram-positive bacteria.
Catalog No. 50-205-3450