Learn More
Abcam Heliquinomycin, Antibiotic agent, 500UG
Catalog No. 502705930
Encompass_Preferred
MW 698.6 Da, Purity >80%. Antibiotic agent. DNA helicase inhibitor (IC₅₀ values are 4.3 and 2.5 μM for DNA helicase B and MCM4/6/7 helicase respectively). Inhibits DNA replication (IC₅₀ values are 1 - 4 μM). Weak topoisomerase I and II inhibitor. Inhibits DNA and RNA synthesis. Does not inhibit protein synthesis. Induces G2/M cell cycle arrest. Shows antibacterial effects. Inhibits cancer cell line growth (IC₅₀ values are 1.4 - 4 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5930 Supplier Abcam Supplier No. AB144268500UG
MW 698.6 Da, Purity >80%. Antibiotic agent. DNA helicase inhibitor (IC₅₀ values are 4.3 and 2.5 μM for DNA helicase B and MCM4/6/7 helicase respectively). Inhibits DNA replication (IC₅₀ values are 1 - 4 μM). Weak topoisomerase I and II inhibitor. Inhibits DNA and RNA synthesis. Does not inhibit protein synthesis. Induces G2/M cell cycle arrest. Shows antibacterial effects. Inhibits cancer cell line growth (IC₅₀ values are 1.4 - 4 μM).
The product is subject to the following: Abcam Restricted Use Statement