Learn More
Waters Corp Waters Corporation Helium Connector Kit, Adapter, with Ferrules
Catalog No. 50793512 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Helium Connector Kit is a spare part kit designed to adapt a helium supply source to a sparge valve inlet bulkhead port.
  • Adapts helium supply source to sparge valve inlet bulkhead port
  • Includes ferrules for solvent inlet tube
Catalog No. 50-793-512 Supplier Waters Corp Supplier No. WAT023486
The Waters Helium Connector Kit is a spare part kit designed to adapt a helium supply source to a sparge valve inlet bulkhead port.
  • Adapts helium supply source to sparge valve inlet bulkhead port
  • Includes ferrules for solvent inlet tube