Learn More
GENERAL BIOPHYSICS LLC Helium Gas Analyzer Model A
Catalog No. NC3361688
Encompass
Thermoconductivity Gas Analyzer for Helium with Onboard PumpApplication: Calibrated for argon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.1%.
Catalog No. NC3361688 Supplier GENERAL BIOPHYSICS LLC Supplier No. GAHEO2A
Add to Cart
Add to Cart
Request a Quote
Thermoconductivity Gas Analyzer for Helium with Onboard PumpApplication: Calibrated for argon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.1%.