Learn More
GENERAL BIOPHYSICS LLC Helium Gas Analyzer Model B
Catalog No. NC3361692
Encompass
Thermoconductivity Gas Analyzer for Helium (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.
Catalog No. NC3361692 Supplier GENERAL BIOPHYSICS LLC Supplier No. GAHEO2B
Thermoconductivity Gas Analyzer for Helium (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.