Learn More
Waters Corp Helium Purifier Kit, for GC-MS Systems, Increases Helium Purity
Catalog No. 502391613 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is used to increase the purity of Helium when it is utilized as a carrier gas in GC-MS systems, including the Xevo TQ-GC system. The kit comprises a Helium Purifier module, Stainless Steel tubing, and the necessary fittings for installation.
  • Increases purity of helium when used as a carrier gas in GC-MS systems.
  • Comprises a helium purifier module.
  • Includes stainless steel tubing.
  • Includes necessary fittings for installation.
  • Suitable for GC-MS systems such as Xevo TQ-GC.
Catalog No. 50-239-1613 Supplier Waters Corp Supplier No. 205002138
This kit is used to increase the purity of Helium when it is utilized as a carrier gas in GC-MS systems, including the Xevo TQ-GC system. The kit comprises a Helium Purifier module, Stainless Steel tubing, and the necessary fittings for installation.
  • Increases purity of helium when used as a carrier gas in GC-MS systems.
  • Comprises a helium purifier module.
  • Includes stainless steel tubing.
  • Includes necessary fittings for installation.
  • Suitable for GC-MS systems such as Xevo TQ-GC.