Learn More
MSC HexArmor Helix 2059 Gloves, A4 Cut, Large, 1 Pair
Catalog No. 5000685116
Encompass
HexArmor Helix 2059 gloves protect hands from cuts, punctures, and abrasion. Featuring durable acrylic and nitrile coating for secure grip and enhanced protection.
  • Knit style for comfort and dexterity
  • Acrylic primary material; nitrile coating for durability
  • Palm and fingertips coated for improved grip
  • Textured grip surface
  • ANSI/ISEA A4 cut, 2 puncture, 4 abrasion resistance
  • Features an 18 gauge construction
  • High-visibility yellow/black design
  • Overall length of 11 in.
Catalog No. 50-006-85116 Supplier MSC Supplier No. 73986978
HexArmor Helix 2059 gloves protect hands from cuts, punctures, and abrasion. Featuring durable acrylic and nitrile coating for secure grip and enhanced protection.
  • Knit style for comfort and dexterity
  • Acrylic primary material; nitrile coating for durability
  • Palm and fingertips coated for improved grip
  • Textured grip surface
  • ANSI/ISEA A4 cut, 2 puncture, 4 abrasion resistance
  • Features an 18 gauge construction
  • High-visibility yellow/black design
  • Overall length of 11 in.