Learn More
HELIX CHROMATOGRAPHY Amaze RP A3 2.1 x 100 mm, 3 um
Supplier:  HELIX CHROMATOGRAPHY AA3211031
Encompass_Preferred
Mixed-mode stationary phase combining reversed-phase, cation-exchange, and anion-exclusion mechanisms. Provides broad retention for amines, hydrophilic and hydrophobic compounds, inorganic cations, basic drugs, and anionic counter-ions. Provides moderate cation-exchange retention with enhanced selectivity for basic analytes, while proprietary ligand design introduces additional chelation interactions for unique separation behavior. Functional groups are integrated on a single ligand. A medium strength acidic group (pKa ~3) is positioned in the inner layer of the stationary phase close to the silica surface, minimizing interaction with residual silanols and improving peak shape for basic analytes. Incorporates surface-bound ion-pairing functionality, enabling full MS compatibility without the need for mobile phase ion-pairing reagents. Provides comparable selectivity and retention to established mixed-mode stationary phases, enabling direct method transfer with minimal adjustment.
Catalog No. NC3977821