Learn More
Glass Expansion Helix CT Locking Screw with Seal
SDP
Catalog No. 501907304
Encompass_Preferred
Helix CT Locking Screw and Seal. A replacement Helix Constant Torque Locking Screw and Seal. This is what secures the nebulizer in Glass Expansion spray chambers and creates the gas tight seal between the nebulizer and the spray chamber. To ensure a gas tight seal replace the seal every 6 months.Damage to the Helix CT insert of a TFE spray chamber can occur over time due to overtightening of the Helix CT locking screw. This will happen if the Helix CT seal is worn and you continue to tighten the locking screw, this results in the nebulizer being pushed further into the spray chamber and the locking screw going past the threads of the Helix insert. Essentially stretching the softer TFE material and slowly deforming the Helix CT insert. The same can occur if the Helix CT locking screw is tightened without the nebulizer in place.To only replace the Helix CT Seal, order NC2081825.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7304 Supplier Glass Expansion Supplier No. 708031439
Helix CT Locking Screw and Seal. A replacement Helix Constant Torque Locking Screw and Seal. This is what secures the nebulizer in Glass Expansion spray chambers and creates the gas tight seal between the nebulizer and the spray chamber. To ensure a gas tight seal replace the seal every 6 months.Damage to the Helix CT insert of a TFE spray chamber can occur over time due to overtightening of the Helix CT locking screw. This will happen if the Helix CT seal is worn and you continue to tighten the locking screw, this results in the nebulizer being pushed further into the spray chamber and the locking screw going past the threads of the Helix insert. Essentially stretching the softer TFE material and slowly deforming the Helix CT insert. The same can occur if the Helix CT locking screw is tightened without the nebulizer in place.To only replace the Helix CT Seal, order NC2081825.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001