Learn More
Microbiologics™ Helix Elite™ Cepheid Xpert™ EV Control Panel
SDP
Catalog No. 23004138 Shop All Microbiologics Products
Change view
Click to view available options
Six positive and negative control swabs
Catalog No.
23-004-138 Six positive and negative control swabs
1 options
This item is not returnable. View return policy
Catalog No. 23-004-138 Supplier Microbiologics™ Supplier No. 8190
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Each kit includes six positive control swabs and six negative control swabs. The positive control consists of Coxsackievirus B1 NCPV 0812141v. The negative control consists of Rhinovirus 1B NCPV 0111101v.
Features the manufacturer recommended QC strains in an easy-to-use format
Specifications
Biosafety Level 1
Description Cepheid Xpert EV Control Panel
Includes Each kit includes six positive control swabs and six negative control swabs. The positive control consists of Coxsackievirus B1 NCPV 0812141v. The negative control consists of Rhinovirus 1B NCPV 0111101v.