Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Helix Elite™ Inactivated SARS-CoV-2 Whole Virus Kit
SDP

Catalog No. 23047378 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5 Swabs
6 Swabs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-378 5 Swabs
23-047-379 6 Swabs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-047-378 Supplier Microbiologics™ Supplier No. HE0065N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Can be used to monitor the extraction, amplification and detection of SARS-CoV-2 molecular applications. Routine use of external quality controls monitor day to day performance of molecular testing procedures.

The Inactivated SARS-CoV-2 Whole Virus is made with whole inactivated SARS-CoV-2 virus (Isolate SARS-CoV-2/USA/WA1/2020) and inactivated A549 lung epithelial cells.

Specifications

Sample Type Virus
Type Control
Includes 5 individually packaged lyophilized pellets and 5 vials of molecular standard water for rehydration
Level 2
Quantity 5 Swabs
Form Pellet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.