Learn More
Microbiologics™ Helix Elite™ Inactivated SARS-CoV-2 Whole Virus Kit
SDP
Catalog No. 23047379 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5 Swabs
6 Swabs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-379 6 Swabs
23-047-378 5 Swabs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-047-379 Supplier Microbiologics™ Supplier No. HE0066NS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Can be used to monitor the extraction, amplification and detection of SARS-CoV-2 molecular applications. Routine use of external quality controls monitor day to day performance of molecular testing procedures.
The Inactivated SARS-CoV-2 Whole Virus is made with whole inactivated SARS-CoV-2 virus (Isolate SARS-CoV-2/USA/WA1/2020) and inactivated A549 lung epithelial cells.
Specifications
Sample Type Swab
Type Control
Includes 6 individually packaged lyophilized swabs
Level 2
Quantity 6 Swabs