Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Helix Elite™ Inactivated SARS-CoV-2 Whole Virus Kit
Shop All Microbiologics Products

Description
The Inactivated SARS-CoV-2 Whole Virus is made with whole inactivated SARS-CoV-2 virus (Isolate SARS-CoV-2/USA/WA1/2020) and inactivated A549 lung epithelial cells.
Specifications
Specifications
| Sample Type | Virus |
| Type | Control |
| Includes | 5 individually packaged lyophilized pellets and 5 vials of molecular standard water for rehydration |
| Level | 2 |
| Quantity | 5 Swabs |
| Form | Pellet |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?