Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Helix Elite™ Inactivated Standard SARS-CoV-2 B.1.1.7 Whole Virus (Alpha Variant)
SDP

Catalog No. 23047412 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5 Pellets (Individually Packaged), 5 Vials of Molecular Standard Water
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-412 5 Pellets (Individually Packaged), 5 Vials of Molecular Standard Water
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-412 Supplier Microbiologics™ Supplier No. HE0071N
This item is not returnable. View return policy

The Inactivated SARS-CoV-2 B.1.1.7 Whole Virus (Alpha Variant) can be used to monitor the extraction, amplification, and detection of SARS-CoV-2 molecular applications. Routine use of external quality controls monitor day to day performance of molecular testing procedures.

  • Fully intact, non-viable process controls
  • Use for every step of the molecular testing process, including extraction, amplification, and detection of SARS-CoV-2
  • Convenient, test-ready format
  • Easy, economical room temperature storage
  • Biosafety Level 1

Specifications

For Use With (Application) Molecular Testing
Product Line Helix Elite™ Inactivated Standard
Quantity 5 Pellets (Individually Packaged), 5 Vials of Molecular Standard Water
Strain Designations SARS-CoV-2 B.1.1.7 (Alpha Variant) Whole Virus
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.