Learn More
Microbiologics™ Helix Elite™ Cepheid Xpert™ CT/NG Control Panel
SDP
Catalog No. 23004129 Shop All Microbiologics Products
Change view
Click to view available options
Six positive and negative controls
Catalog No.
23-004-129 Six positive and negative controls
1 options
This item is not returnable. View return policy
Catalog No. 23-004-129 Supplier Microbiologics™ Supplier No. 8188
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Each kit includes six positive control swabs and six negative control swabs. The positive control includes Chlamydia trachomatis (strain LGV-2-434), Neisseria gonorrhoeae and human cells. The negative control includes human cells.
Features the manufacturer recommended QC strains in an easy-to-use format
Specifications
Biosafety Level 1
Includes Six positive control swabs and six negative control swabs. The positive control includes Chlamydia trachomatis (strain LGV-2-434), Neisseria gonorrhoeae and human cells. The negative control includes human cells.