Learn More
Glass Expansion Helix Seal (Pkt. 4)
SDP
Catalog No. NC1417609
Encompass_Preferred
Helix Seal (PKT 4). Package of four replacement Helix Seals. In conjunction with the Locking Screw, this is what creates the gas tight seal between the nebulizer and the spray chamber. To ensure a proper seal replace every 6 months.Damage to the Helix insert of a TFE spray chamber can occur over time due to overtightening of the Helix locking screw. This will happen if the Helix seal is worn and you continue to tighten the locking screw, this results in the nebulizer being pushed further into the spray chamber and the locking screw going past the threads of the Helix insert. Essentially stretching the softer TFE material and slowly deforming the Helix insert. The same can occur if the Helix locking screw is tightened without the nebulizer in place.To only replace the Helix Locking Screw, order NC1450039.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC1417609 Supplier Glass Expansion Supplier No. 701000237
Helix Seal (PKT 4). Package of four replacement Helix Seals. In conjunction with the Locking Screw, this is what creates the gas tight seal between the nebulizer and the spray chamber. To ensure a proper seal replace every 6 months.Damage to the Helix insert of a TFE spray chamber can occur over time due to overtightening of the Helix locking screw. This will happen if the Helix seal is worn and you continue to tighten the locking screw, this results in the nebulizer being pushed further into the spray chamber and the locking screw going past the threads of the Helix insert. Essentially stretching the softer TFE material and slowly deforming the Helix insert. The same can occur if the Helix locking screw is tightened without the nebulizer in place.To only replace the Helix Locking Screw, order NC1450039.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001