Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cramer Helix Stool

Catalog No. 03155055
Change view
Click to view available options
Back Style:
Ergonomic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Back Style
03-155-055 Ergonomic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-155-055 Supplier Cramer Supplier No. HXHU12L
Add to Cart
Add to Cart

Helix keeps pace with a Frameless Back™ that flexes naturally and adapts instantly. It’s seating that stays out of the way, so focus stays on the work.

Frameless Back™ Patent-pending Frameless Back™ adapts organically to the user, providing exceptional freedom of motion and comfort. i+™ Polyurethane Seamless i+™ Polyurethane cushions resist contaminants and hold up to hospital cleaning agents. Complies with BIFMA Healthcare Furniture Cleanability Guidelines HCF 8.1-2019. Welcome to the future of seating for labs and caregivers. One-touch adjustment One-touch adjustment ensures a fast, personal fit for multiple users. Modern Aesthetic Unique curves and fresh optional color accents offer a calming, modern aesthetic.

Features

  • Patent-pending Frameless Back™ adapts organically to the user, providing exceptional freedom of motion and comfort
  • One-touch adjustment ensures a fast, personal fit for multiple users
  • Seamless i+™ Polyurethane cushions resist contaminants and will withstand hospital cleaning agents
  • Streamlined form minimizes the visual impact and doesn't interrupt sightlines
  • Meets BIFMA Healthcare Furniture Cleanability Guidelines HCF 8.1-2019
  • ISO-Class 4 cleanroom rated
  • 300 lb. weight capacity
  • SCS Indoor Advantage Gold Certification and may help contribute toward LEED certification

Construction

  • i+™ Polyurethane seat and back
  • Advanced thermoplastic elastomer flexing upright
  • Heavy-duty formed steel mechanism
  • Resilient glass fiber support spring
  • 27.75 in. durable nylon base
  • Class 4 pneumatic cylinder

Options

  • Fixed arm option available
  • High-performance twist-lock flat surface steel 22 in. black footring. Footring is only available on high-height
  • Casters: multi-surface dual wheel is standard. Non-rolling glides and breaking casters are also available
  • Desk and high-height with desk to counter option

Specifications

Product Type High Stool
With Lumbar Support No
With Tilt No
Footrest Yes
With Casters Yes
With Armrest No
With Backrest Yes
Color Black, Gray
Back Style Ergonomic
Material Polypropylene
Seating Type Stool with Back
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.