Learn More
Helix Medical Inc HELIX TUB P SIL .125X.250 50FT
Catalog No. NC0842615
Encompass_Preferred
Medical-grade tubing is ideal for use in the healthcare industry. No leaching of volatile by-products, bloom or peroxide residues. Autoclavable. Coil length: 50'. I.D. x O.D. : 0.125 x 0.250" Wall Thickness : 0.063"
Catalog No. NC0842615 Supplier Helix Medical Inc Supplier No. 6080521
Medical-grade tubing is ideal for use in the healthcare industry. No leaching of volatile by-products, bloom or peroxide residues. Autoclavable. Coil length: 50'. I.D. x O.D. : 0.125 x 0.250" Wall Thickness : 0.063"