Learn More
Molecular Dimensions Helix HT-96, 96 x 1 mL
SDP
Catalog No. NC1029490
Encompass
Crystallize a diverse range of nucleic acids, oligonucleotides, triplexes, quadruplexes, pseudoknots, i-motifs, and large molecular weight nucleic acids.
Catalog No. NC1029490 Supplier Molecular Dimensions Supplier No. MD169
Crystallize a diverse range of nucleic acids, oligonucleotides, triplexes, quadruplexes, pseudoknots, i-motifs, and large molecular weight nucleic acids.