Learn More
Alkali Scientific Hellendahl Glass Staining Jar with Glass Lid, 9 Slide Capacity, for 25×75 mm Slides, 20/Case
SDP
Catalog No. 5000662765
Encompass_Preferred
This Hellendahl type glass staining jar is designed to accommodate up to 9 standard 25.0 × 75.0 mm (1" × 3") microscope slides for efficient staining in histology and pathology laboratories. The vertical slide arrangement allows consistent reagent contact across multiple slides while keeping samples organized during processing. Manufactured from durable laboratory grade glass, the jar provides excellent chemical resistance and clear visibility of contents. A matching glass lid is included to help reduce evaporation and protect staining solutions from contamination during use. The jar is suitable for routine slide staining procedures and laboratory workflows requiring reliable reagent exposure.
Catalog No. 50-006-62765 Supplier Alkali Scientific Supplier No. SJ009CS
This Hellendahl type glass staining jar is designed to accommodate up to 9 standard 25.0 × 75.0 mm (1" × 3") microscope slides for efficient staining in histology and pathology laboratories. The vertical slide arrangement allows consistent reagent contact across multiple slides while keeping samples organized during processing. Manufactured from durable laboratory grade glass, the jar provides excellent chemical resistance and clear visibility of contents. A matching glass lid is included to help reduce evaporation and protect staining solutions from contamination during use. The jar is suitable for routine slide staining procedures and laboratory workflows requiring reliable reagent exposure.