Learn More
Electron Microscopy Sciences Hellendahl Staining Dish 1 each
SDP
Catalog No. 5029861
Encompass_Preferred
The Hellendahl type dish for the staining or use as a TLC developing chamber. Holds up to 8 individual 3" x 1" (76 x 26 mm) slides, or 16 slides back-to-back. These dishes are made from soda lime glass. The approximate inside dimensions: 76 mm long x 25 mm wide x 75 mm deep.
Catalog No. 50-298-61 Supplier Electron Microscopy Sciences Supplier No. 7031301
The Hellendahl type dish for the staining or use as a TLC developing chamber. Holds up to 8 individual 3" x 1" (76 x 26 mm) slides, or 16 slides back-to-back. These dishes are made from soda lime glass. The approximate inside dimensions: 76 mm long x 25 mm wide x 75 mm deep.