Learn More
Bioworld Heller Medium, 50 L
Catalog No. 50254620
Encompass_Preferred
Heller MediumContains Macronutrients and Micronutrients as described by Heller (1953) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-620 Supplier Bioworld Supplier No. 306300363
Heller MediumContains Macronutrients and Micronutrients as described by Heller (1953) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested