Learn More
MCR Safety HellKat 3 Foam Lined MAX6 Lens
Catalog No. 19812694 Shop All MCR Safety Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-812-694 Clear
19-812-699 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-812-694 Supplier MCR Safety Supplier No. HK310PF
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
HellKat 3 Foam Lined MAX6 Lens gives you all the features you want for a comfortable and stylish option in protective eyewear. This generous lens offers an outstanding orbital seal and prevents hazards from threatening one of your most treasured belongings - your eyesight.
  • Lightweight, contoured frameless design
  • MAX6™ premium anti-fog lens coating
  • Excellent orbital seal for eye protection
  • Non-slip soft bayonet temple
  • Soft, secure TPR nose pad
  • HellKat® 3 offers closed cell foam lining
Specifications
Lens Tint Clear
For Use With (Application) Manufacturing
Frame Color Clear
No. per Pack 1 Pair
Lens Material Polycarbonate
Frame Material Polycarbonate