Learn More
Sigma Aldrich Fine Chemicals Biosciences Hellma(R) absorption cuvettes, semi Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 1,400 muL | 1EA
Catalog No. NC1161420
Encompass
Hellma(R) absorption cuvettes, semi Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 1,400 muL | 1EA
Catalog No. NC1161420 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6002881EA
Hellma(R) absorption cuvettes, semi Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 1,400 muL | 1EA