Learn More
Sigma Aldrich Fine Chemicals Biosciences HELLMA ABSORPTION CUVETTE, STD
Catalog No. NC1811770
Encompass
optical glass, spectral range 360-2,500 nm, pathlength 10 mm, chamber volume 3,500 μL
Catalog No. NC1811770 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6000081EA
optical glass, spectral range 360-2,500 nm, pathlength 10 mm, chamber volume 3,500 μL