Learn More
Sigma Aldrich Fine Chemicals Biosciences Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 400 muL | 1EA
Catalog No. NC2231102
Encompass
Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 400 muL | 1EA
This item is not returnable. View return policy
Catalog No. NC2231102 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6008221EA
Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 400 muL | 1EA