Learn More
Sigma Aldrich Fine Chemicals Biosciences Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 700 muL | 1EA
Catalog No. NC2231105
Encompass
Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 700 muL | 1EA
Catalog No. NC2231105 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6002101EA
Hellma(R) absorption cuvettes, Micro Suprasil(R) quartz, spectral range 200-2,500 nm, pathlength 10 mm, chamber volume 700 muL | 1EA