Learn More
Sigma Aldrich Fine Chemicals Biosciences HellmanexTM III Special Cl1EA
Catalog No. 502900305
Encompass
Hellmanex III significantly reduces the surface tension of water. The removal of dirt particles is also assured by the good wetting action of a Hellmanex III aqueous SOLUTIONon while its high emulsifying and dispersing capabilities prevent the redeposition of the loosened particles. Special surface-active substances facilitate the residue-free rinsing of the optical components once they have been cleaned.Hellmanex III is filtered to 1 mum and is therefore virtually free of solid particles.
Catalog No. 50-290-0305 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z8059391EA
Hellmanex III significantly reduces the surface tension of water. The removal of dirt particles is also assured by the good wetting action of a Hellmanex III aqueous SOLUTIONon while its high emulsifying and dispersing capabilities prevent the redeposition of the loosened particles. Special surface-active substances facilitate the residue-free rinsing of the optical components once they have been cleaned.Hellmanex III is filtered to 1 mum and is therefore virtually free of solid particles.