Learn More
Sigma Aldrich Fine Chemicals Biosciences HellmaR absorption cuvette1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6269291EA
Encompass
flowthrough cells with 2 screw connectors M 6 X 1 and FEP tubes aperture 17.5mm x 3.5mm rectangular
Catalog No. 50-290-8572