Learn More
Sigma Aldrich Fine Chemicals Biosciences HellmaR fluorescence cuvet1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z6007171EA
Encompass
Hellma(R) fluorescence cuvettes standard cells Macro with PTfE stoppers (spectral range 200-2 500 nm pathlength 10x10 mm chamber volume 3 500 muL) are available in high-performance Suprasil quartz material. The highest level of sensitivity in the measurement can be achieved by suitably selecting the cell for a specific application such that it exhibits high transmission in the spectral range of interest. The maximum of 80-90% transmission shown by empty Suprasil quartz cell is caused by the reflection loss at the four glass/air boundaries. When filled with common solvents the cell will exhibit significantly higher maximum transmissions.
Catalog No. 50-290-5984
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.