Learn More
MSC Helly Hansen Rain Jacket, PVC, Green, Medium, 1 ea
Catalog No. 5000792369
Encompass
This rain jacket is designed for demanding outdoor conditions, offering robust protection against water, oil, and chemicals. Its durable construction ensures reliability for professionals in various challenging environments.
  • Keeps water out
  • Heavy-weight design
  • Double storm flap with snap closures
  • Drawcord hood
  • Extra durable construction
  • Oil-resistant
  • Chemical-resistant
  • Cold-resistant coating and fabric
  • Highly waterproof
  • Designed for layering without restricting movement
  • Made from PVC-coated woven cotton
  • EN343 certified
Catalog No. 50-007-92369 Supplier MSC Supplier No. 81735664
This rain jacket is designed for demanding outdoor conditions, offering robust protection against water, oil, and chemicals. Its durable construction ensures reliability for professionals in various challenging environments.
  • Keeps water out
  • Heavy-weight design
  • Double storm flap with snap closures
  • Drawcord hood
  • Extra durable construction
  • Oil-resistant
  • Chemical-resistant
  • Cold-resistant coating and fabric
  • Highly waterproof
  • Designed for layering without restricting movement
  • Made from PVC-coated woven cotton
  • EN343 certified