Learn More
MSC Helly Hansen Rain Jacket, Small, Yellow, Polyester, Waterproof, 1 ea
Catalog No. 5000796826
Encompass
This rain jacket is constructed from polyester and designed for protection in wet conditions. It features a storm flap and is waterproof, making it suitable for various outdoor applications.
  • Waterproof design
  • Equipped with a storm flap
  • Certified with ASTM F1671 and EN343
  • Features snap closure for secure fitting
  • Includes 2 functional pockets
  • Suitable for chest sizes 34 to 36 inch
Catalog No. 50-007-96826 Supplier MSC Supplier No. 09221441
This rain jacket is constructed from polyester and designed for protection in wet conditions. It features a storm flap and is waterproof, making it suitable for various outdoor applications.
  • Waterproof design
  • Equipped with a storm flap
  • Certified with ASTM F1671 and EN343
  • Features snap closure for secure fitting
  • Includes 2 functional pockets
  • Suitable for chest sizes 34 to 36 inch