Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanSpace™ Technology Helmet Hook Strap Accessory

Catalog No. 17800623 Shop All Cleanspace Technology Products
Change view
Click to view available options
For Use With (Equipment):
PAF-1014, PAF-1106
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-800-623 PAF-1014, PAF-1106
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-800-623 Supplier CleanSpace™ Technology Supplier No. PAF0075
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Helmet hook strap accessory is designed for use when wearing the respirator with a safety helmet.

  • Used with the revolutionary CleanSpace™ Respirator: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement

Specifications

Disposable Reusable
Product Type Helmet Hook Strap
For Use With (Application) Welding, Woodworking, Manufacturing, Smelting, Construction, Recycling Plants, Emergency Services, Mining, Agriculture, Processing Plants, Grinding
For Use With (Equipment) PAF-1014, PAF-1106
Model PAF-0075
Storage Requirements Store at -10°C to +55°C
Dimensions (L x W x H) 950 x 350 x 600 mm
Weight (Metric) 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.