Learn More
Aviva Systems Biology Corp HELT Antibody - middle region (ARP50404_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50404_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HELT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HELT. Alternate names: HCM1228, HESL, Mgn, bHLHb44ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53630