Learn More
AdipoGen Helvolic acid
SDP
Catalog No. 502561931
Encompass_Preferred
Chemical. CAS 29400-42-8. Formula C33H44O8. MW 568.7. Isolated from Aspergillus fumigatus Strain WDMH-35. Steroidal triterpene. Antibiotic. Antibacterial. Shows broad activity against Gram positive and negative bacteria. Mycotoxin. Antifungal. Antitumor agent. Antituberculosis agent. Oxidized LDL uptake inhibitor. Phytotoxin. Inhibitor of the protein biosynthesis through archeal elongation factor 2 EF-2.
Catalog No. 50-256-1931 Supplier AdipoGen Supplier No. BVT0435M001
Chemical. CAS 29400-42-8. Formula C33H44O8. MW 568.7. Isolated from Aspergillus fumigatus Strain WDMH-35. Steroidal triterpene. Antibiotic. Antibacterial. Shows broad activity against Gram positive and negative bacteria. Mycotoxin. Antifungal. Antitumor agent. Antituberculosis agent. Oxidized LDL uptake inhibitor. Phytotoxin. Inhibitor of the protein biosynthesis through archeal elongation factor 2 EF-2.