Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems ACROBiosystems Influenza A (A/Darwin/6/2021) H3N2 Hemagglutinin (HA) Protein, His-tagged, Trimeric, Lyophilized, 100 μg

Catalog No. 502357291
Encompass_Preferred

ACROBiosystems recombinant Influenza A hemagglutinin (HA) protein from the A/Darwin/6/2021 (H3N2) strain is supplied as a C-terminal His-tagged, trimeric construct expressed in HEK293 cells and provided lyophilized for long-term storage.

  • Expressed in human HEK293 cells and designed as a trimer.
  • Lyophilized preparation with trehalose protectant for -20°C storage.
  • Calculated molecular weight 63 kDa; migrates 70-90 kDa on reducing SDS-PAGE due to glycosylation.
  • SEC-MALS shows trimer mass around 225-255 kDa.
  • Purity greater than 90% by SDS-PAGE and by SEC-MALS.
  • Endotoxin less than 1.0 EU/μg (LAL / rFC methods).
  • Contains a C-terminal polyhistidine tag for capture or detection.
  • QC-tested for ELISA binding to anti-HA (H3N2) antibody.

Catalog No. 50-235-7291 Supplier ACROBiosystems Supplier No. HA2V52H5100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACROBiosystems recombinant Influenza A hemagglutinin (HA) protein from the A/Darwin/6/2021 (H3N2) strain is supplied as a C-terminal His-tagged, trimeric construct expressed in HEK293 cells and provided lyophilized for long-term storage.

  • Expressed in human HEK293 cells and designed as a trimer.
  • Lyophilized preparation with trehalose protectant for -20°C storage.
  • Calculated molecular weight 63 kDa; migrates 70-90 kDa on reducing SDS-PAGE due to glycosylation.
  • SEC-MALS shows trimer mass around 225-255 kDa.
  • Purity greater than 90% by SDS-PAGE and by SEC-MALS.
  • Endotoxin less than 1.0 EU/μg (LAL / rFC methods).
  • Contains a C-terminal polyhistidine tag for capture or detection.
  • QC-tested for ELISA binding to anti-HA (H3N2) antibody.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.