Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Influenza A (Sydney/5/2021) Hemagglutinin (HA) Protein, His-Tagged, 100 μg

Catalog No. 502370251
Encompass_Preferred

Recombinant Influenza A (Sydney/5/2021, H1N1) hemagglutinin (HA) protein produced in HEK293 cells, supplied lyophilized with a C-terminal His tag. Verified by SEC-MALS and suitable for binding and bioactivity assays such as ELISA and SPR.

  • Lyophilized powder; 100 μg.
  • C-terminal polyhistidine tag.
  • Produced in human 293 (HEK293) expression system.
  • Calculated molecular weight 63.1 kDa; migrates 90-110 kDa on SDS-PAGE; trimer ~215-245 kDa by SEC-MALS.
  • Purity greater than 90% by SDS-PAGE and SEC-MALS.
  • Storage: -20°C or lower (lyophilized); -70°C up to 3 months after reconstitution.
  • Formulation: 0.1 M sodium citrate, pH 5.5, lyophilized with trehalose as protectant.
  • Validated for bioactivity in ELISA and SPR assays.

Catalog No. 50-237-0251 Supplier ACROBiosystems Supplier No. HA1V52H4100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Influenza A (Sydney/5/2021, H1N1) hemagglutinin (HA) protein produced in HEK293 cells, supplied lyophilized with a C-terminal His tag. Verified by SEC-MALS and suitable for binding and bioactivity assays such as ELISA and SPR.

  • Lyophilized powder; 100 μg.
  • C-terminal polyhistidine tag.
  • Produced in human 293 (HEK293) expression system.
  • Calculated molecular weight 63.1 kDa; migrates 90-110 kDa on SDS-PAGE; trimer ~215-245 kDa by SEC-MALS.
  • Purity greater than 90% by SDS-PAGE and SEC-MALS.
  • Storage: -20°C or lower (lyophilized); -70°C up to 3 months after reconstitution.
  • Formulation: 0.1 M sodium citrate, pH 5.5, lyophilized with trehalose as protectant.
  • Validated for bioactivity in ELISA and SPR assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.