Learn More
StatLab™ Hema-Diff 1 Fixative
Description
- Entire procedure takes 30 seconds
- Air dried specimens are fixed in Solution I, then dipped in Solutions II and III
- Can also be used as a special stain for Helicobacter pylori
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Application) | Differential Staining |
| Quantity | 1 gal. |
| Product Type | Fixative |