Learn More
StatLab™ Hema-Diff 2 Xanthene
GSA_VASDP
Catalog No. STL1197516
Change view
Click to view available options
Quantity:
16 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL1197516 16 oz.
STL119751 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL1197516 Supplier StatLab™ Supplier No. 1197516
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
A three-step procedure for differential staining of blood smears that is used extensively in FNA cytology.
  • Entire procedure takes 30 seconds
  • Air dried specimens are fixed in Solution I, then dipped in Solutions II and III
  • Can also be used as a special stain for Helicobacter pylori
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Application) Differential Staining
Quantity 16 oz.
Product Type Xanthene